Learn More
Obeticholic acid, MedChemExpress
Código de producto. 16491267
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
200 miligramos
5 miligramos
50 miligramos
500 miligramos
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
16491267 50 mg 50 miligramos
16477757 5 mg 5 miligramos
16475776 100 mg 100 miligramos
16431036 200 mg 200 miligramos
16415038 10 mg 10 miligramos
16481626 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Código de producto. 16491267 Proveedor Medchem Express N.º de proveedor HY12222
MedChemExpress Obeticholic acid (INT-747) is a potent, selective and orally active FXR agonist with an EC50 of 99 nM. Obeticholic acid has anticholeretic and anti-inflammation effect. Obeticholic acid also induces autophagy.
Obeticholic acid (INT-747) is a potent, selective and orally active FXR agonist with an EC50 of 99 nM. Obeticholic acid has anticholeretic and anti-inflammation effect. Obeticholic acid also induces autophagy.
Especificaciones
Nombre del producto químico o material Obeticholic acid
CAS 459789-99-2
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C26H44O4
Cantidad 50 mg
Sinónimo INT-747 6-ECDCA 6-Ethylchenodeoxycholic acid
Información de solubilidad DMSO : ≥ 100 mg/mL (237.74 mM) ∣Ethanol : ≥ 50 mg/mL (118.87 mM)
SMILES C[C@@]([C@]1([H])[C@@H](CC)[C@H]2O)(CC[C@@H](O)C1)[C@]3([H])[C@]2([H])[C@@](CC[C@]4([H])[C@H](C)CCC(O)=O)([H])[C@]4(C)CC3
Peso molecular (g/mol) 420.63
Formula Weight (peso de la fórmula) 420.63
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid