Learn More
STEMCELL Technologies OAC1
Description
- OAC1 is an OCT-4-activating compound that activates expression through the OCT-4 gene promoter (Li et al.) Oct-4 (POU5F1) is a transcription factor that is critically involved in the self-renewal of pluripotent stem cells, and its expression is commonly used as a marker for pluripotency
- Enhances the reprogramming efficiency of mouse embryonic fibroblasts transfected with OCT4, SOX2, KLF4, and c-MYC (Li et al.)
- Mediates ex vivo expansion of cord blood CD34+ hematopoietic stem and progenitor cells (Huang et al.)
Specifications
Specifications
| CAS | 300586-90-7 |
| Cantidad | 5 mg |
| Fórmula molecular | C14H11N3O |
| Pureza | ≥98% |