Learn More
Tocris Bioscience™ O4I2
Oct3/4 inducer; induces expression of pluripotent-associated genes
Marca: Tocris Bioscience™ 5664/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
O4I2 is a Oct3/4 inducer; promotes Oct3/4 expression and stabilizes the Oct3/4 protein in HEK293 cells. Induces expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Induces 15-fold increase in Oct3/4 expression in human primary fibroblasts.Especificaciones
| Ethyl 2-[(4-chlorophenyl)amino]-4-thiazolecarboxylate | |
| 165682-93-9 | |
| 50 mg |