Learn More
NVX-207, MedChemExpress
Código de producto. 16463448
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16463448 10 mg 10 miligramos
16434656 100 mg 100 miligramos
16419956 50 mg 50 miligramos
16416177 5 mg 5 miligramos
16402776 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16463448 Proveedor Medchem Express N.º de proveedor HY101597
MedChemExpress NVX-207, a Betulinic acid-derived anti-cancer compound, shows anti-tumor activity (mean IC50=3.5 μM) against various human and canine cell lines. NVX-207-induced apoptosis is associated with activation of the intrinsic apoptotic pathway via cleavage of caspases -9, -3, -7 and of PARP.
NVX-207, a Betulinic acid-derived anti-cancer compound, shows anti-tumor activity (mean IC50=3.5 μM) against various human and canine cell lines. NVX-207-induced apoptosis is associated with activation of the intrinsic apoptotic pathway via cleavage of caspases -9, -3, -7 and of PARP.
Especificaciones
Nombre del producto químico o material NVX-207
Forma física Solid
CAS 745020-66-0
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C36H59NO6
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : 125 mg/mL (207.69 mM; Need ultrasonic)
SMILES O=C([C@]1(CC[C@H]2C(C)=C)[C@@]2([H])[C@](CC[C@@]3([H])[C@]4(CC[C@]5([H])[C@@]3(CC[C@H](OC(C)=O)C5(C)C)C)C)([H])[C@@]4(C)CC1)OCC(CO)(N)CO
Peso molecular (g/mol) 601.86
Formula Weight (peso de la fórmula) 601.86
Porcentaje de pureza 98.0%