missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP88 Polyclonal Antibody, Invitrogen™
Descripción
The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nup88 (nucleoporin 88 kDa) is a 741 amino acid protein that localizes to the nucleus and functions as an essential component of the nuclear pore complex. Expressed ubiquitously, Nup88 is subject to phosphorylation by ATM or ATR and is upregulated in malignant neoplasms and precancerous dysplasias, suggesting a role in tumorigenesis. The gene encoding Nup88 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes.
Especificaciones
Especificaciones
| Antígeno | NUP88 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| génica | NUP88 |
| N.º de referencia del gen | Q8CEC0, Q99567 |
| Alias de gen | 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; nuclear pore complex protein Nup88; nucleoporin 88; nucleoporin 88kDa; nucleoporin Nup84; Nucleoporin Nup88; Nup84; NUP88; Prei2; preimplantation protein 2 |
| Símbolos de los genes | NUP88 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?