Learn More
Thermo Scientific™ Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Descripción
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc Microwell 96-Well Microplates with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
Especificaciones
Especificaciones
| Código de barras | No Barcode |
| Tapa | Con tapa |
| matriz, red, conjunto | 8 x 12 |
| Línea de productos | Nunc |
| Color | Transparente |
| Desechables | Single-use |
| Tipo de producto | Microplate |
| Material | Polystyrene |
| N.° de pocillos | 96 |
| Esterilidad | Estéril |
| Mostrar más |
Para uso exclusivo en investigación.