Learn More
Thermo Scientific™ NUCLEIC-CARD™ matrix, 4 spots
Código de producto. 15692406
Change view
Click to view available options
Cantidad:
100 tarjetas
Tamaño de la unidad:
paquete de 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
15692406 100 tarjetas paquete de 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 15692406 Proveedor Thermo Scientific™ N.º de proveedor 4473977
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
NUCLEIC-CARD is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature.
The NUCLEIC-CARD matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD collection devices are available in one-, two-, and four-spot formats. They are manufactured by Copan, a market leader in the area of sample collection tools, and distributed by Thermo Fisher Scientific.
Key Features:
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms, enabling long-term room temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
For Forensic or Paternity Use Only.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento 100 NUCLEIC-CARD 4 Spots
Store (at room temperature)
Formato Card
Para utilizar con (equipo) Harris Manual Punching System, BSD Robotics Punching System
Línea de productos NUCLEIC-CARD
Cantidad 100 tarjetas
Condiciones de envío Temperatura ambiente