Learn More
Thermo Scientific™ NUCLEIC-CARD™ matrix, 4 spots
Código de producto. 15692406
Change view
Click to view available options
Cantidad:
100 tarjetas
Tamaño de la unidad:
paquete de 100
15692406 100 tarjetas paquete de 100
1 options
Código de producto. 15692406 Proveedor Thermo Scientific™ N.º de proveedor 4473977
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
NUCLEIC-CARD is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature.
The NUCLEIC-CARD matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD collection devices are available in one-, two-, and four-spot formats. They are manufactured by Copan, a market leader in the area of sample collection tools, and distributed by Thermo Fisher Scientific.
Key Features:
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms, enabling long-term room temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
For Forensic or Paternity Use Only.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento 100 NUCLEIC-CARD 4 Spots
Store (at room temperature)
Formato Card
Para utilizar con (equipo) Harris Manual Punching System, BSD Robotics Punching System
Línea de productos NUCLEIC-CARD
Cantidad 100 tarjetas
Condiciones de envío Temperatura ambiente