Learn More
Thermo Scientific™ NUCLEIC-CARD™ matrix, 1 spot
Descripción
The NUCLEIC-CARD matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD collection devices are available in one-, two-, and four-spot formats. They are manufactured by Copan, a market leader in the area of sample collection tools, and distributed by Thermo Fisher Scientific.
Key Features:
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms, enabling long-term room temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
For Forensic or Paternity Use Only.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
Especificaciones
Especificaciones
| Contenido y almacenamiento | 100 NUCLEIC-CARD 1 Spot Store (at room temperature) |
| Formato | Card |
| Para utilizar con (equipo) | Harris Manual Punching System, BSD Robotics Punching System |
| Línea de productos | NUCLEIC-CARD |
| Cantidad | 100 tarjetas |
| Condiciones de envío | Temperatura ambiente |