Learn More
NU 2058, Tocris Bioscience™
Código de producto. 18064552
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18064552 10 mg 10 miligramos
18086022 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18064552 Proveedor Tocris Bioscience N.º de proveedor 3135/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Cdk1 and cdk2 inhibitor
Cyclin-dependent kinase (cdk) 1 and cdk2 inhibitor (Ki values are 5 and 12 μM respectively). Inhibits growth of human tumor cells in vitro (mean GI50 = 13 μM). Also inhibits DNA topoisomerase II ATPase activity (IC50 = 300 μM).
TRUSTED_SUSTAINABILITY
CAS 161058-83-9
Fórmula molecular C12H17N5O
Peso molecular (g/mol) 247.302
Clave InChI MWGXGTJJAOZBNW-UHFFFAOYSA-N
Sinónimo 6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
PubChem CID 4564
Nombre IUPAC 6-(cyclohexylmethoxy)-7H-purin-2-amine
SMILES C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N
Especificaciones
Nombre del producto químico o material NU 2058
Cantidad 10 mg
Formula Weight (peso de la fórmula) 247.3
Porcentaje de pureza >99%