Learn More
NSF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16850785 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16850785 20 μL 20 microlitros
16840785 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16850785 Proveedor Proteintech N.º de proveedor 211721AP20UL
Rabbit Polyclonal Antibody
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
Especificaciones
Antígeno NSF
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.38 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica NSF
N.º de referencia del gen P46459, P46460
Alias de gen NEM sensitive fusion protein, NSF, SKD2, Vesicle fusing ATPase, Vesicular fusion protein NSF
Símbolos de los genes Nsf
Especie del huésped Rabbit
Inmunógeno NSF Fusion Protein Ag15422
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18195, 4905
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG