Learn More
NS-398, MedChemExpress
Código de producto. 16492057
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16492057 50 mg 50 miligramos
16468547 5 mg 5 miligramos
16456356 100 mg 100 miligramos
16405918 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16492057 Proveedor Medchem Express N.º de proveedor HY13913
MedChemExpress NS-398 is a non-steroidal an-inflammatory agent with analgesic and antipyretic effects, and selectively inhibits prostaglandin G/H synthase 2/cyclooxygenase 2 (COX-2) activity, with an IC50 of 3.8 μM, and has no effect on COX-1 at 100 μM.
NS-398 is a non-steroidal an-inflammatory agent with analgesic and antipyretic effects, and selectively inhibits prostaglandin G/H synthase 2/cyclooxygenase 2 (COX-2) activity, with an IC50 of 3.8 μM, and has no effect on COX-1 at 100 μM.
Especificaciones
Nombre del producto químico o material NS-398
CAS 123653-11-2
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C13H18N2O5S
Cantidad 50 mg
Información de solubilidad DMSO : 100 mg/mL (318.11 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC=C([N+]([O-])=O)C=C1OC2CCCCC2)=O
Peso molecular (g/mol) 314.36
Formula Weight (peso de la fórmula) 314.36
Porcentaje de pureza 98.7%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation