Learn More
Tocris Bioscience™ NS 398
Cyclooxygenase-2 (COX-2) inhibitor
Marca: Tocris Bioscience™ 0942/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NS 398 is a selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.Especificaciones
| N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide | |
| >99% | |
| Soluble to 100mM in DMSO |
| 123653-11-2 | |
| 10 mg |
Título del producto
For Research Use Only