missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOTCH2 Chimeric Recombinant Mouse Monoclonal Antibody (B9)

Descripción
Specificity: An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2 but not NRR1.
Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
Especificaciones
Especificaciones
| Antígeno | NOTCH2 Chimeric |
| Aplicaciones | ELISA, Inhibition Assays, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | B9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% ProClin 300 |
| génica | NOTCH2 |
| N.º de referencia del gen | O35516, Q04721 |
| Alias de gen | AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?