Learn More
NKL 22, MedChemExpress
Código de producto. 16499546
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16499546 50 mg 50 miligramos
16492978 10 mg 10 miligramos
16454296 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16499546 Supplier Medchem Express Supplier No. HY100384
This item is not returnable. View return policy
MedChemExpress NKL 22 (compound 4b) is a potent and selective inhibitor of histone deacetylases (HDAC), with an IC50 of 199 and 69 nM for HDAC1 and HDAC3, respectively. NKL 22 exhibits selectivity over HDAC2/4/5/7/8 (IC50≥1.59 μM). NKL 22 ameliorates the disease phenotype and transcriptional abnormalities in Huntington's disease transgenic mice.
NKL 22 (compound 4b) is a potent and selective inhibitor of histone deacetylases (HDAC), with an IC50 of 199 and 69 nM for HDAC1 and HDAC3, respectively. NKL 22 exhibits selectivity over HDAC2/4/5/7/8 (IC50≥1.59 μM). NKL 22 ameliorates the disease phenotype and transcriptional abnormalities in Huntington's disease transgenic mice.
Specifications
Nombre del producto químico o material NKL 22
Forma física Solid
CAS 537034-15-4
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C19H23N3O2
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 29 mg/mL (89.12 mM)
SMILES O=C(NC1=CC=CC=C1N)CCCCCC(NC2=CC=CC=C2)=O
Peso molecular (g/mol) 325.4
Formula Weight (peso de la fórmula) 325.4
Porcentaje de pureza 97.27%