Learn More
Tocris Bioscience™ NK 252
Nrf2 activator
Marca: Tocris Bioscience™ 5293/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NK 252 is a Nrf2 activator; interacts with the Nrf2-binding site of Keap1. Downregulates the expression of fibrogenic genes and increases antioxidant effects in NASH rat models of liver disease.Especificaciones
| NK 252 | |
| N-[5-(2-Furanyl)-1,3,4-oxadiazol-2-yl]-N'-(2-pyridinylmethyl)urea | |
| Soluble to 100mM in 1eq. HCl and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 1414963-82-8 | |
| 50 mg | |
| Powder |
Título del producto
Research Use Only. Not For Human or Veterinary Use