missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584738
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: LVCNGLIHGI ASFVRGGCAS GLYPDAFAPV AQFVNWIDSI IQRSEDNPCP HP Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 65%.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
Especificaciones
| Neutrophil elastase | |
| Polyclonal | |
| Unconjugated | |
| ELANE | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08246 | |
| ELANE | |
| Recombinant protein corresponding to Human ELANE. Recombinant protein control fragment (Product #RP-107524). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido