Learn More
Tocris Bioscience™ Neuropeptide S (Mouse)
Descripción
Neuropeptide S (Mouse) is a potent endogenous neuropeptide S receptor (NPSR) agonist (EC50 = 3 nM). Induces mobilization of intracellular Ca2+. Increases locomotor activity and wakefulness in mice. Also reduces anxiety-like behavior in mice.
Especificaciones
Especificaciones
| CAS | 412938-74-0 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | Additional Peptide Receptor Products |
| Peso molecular | Observed M.W.: 2182.47 |
| Cantidad | 1 mg |
| Forma estructural | C93H156N34O27 |