Learn More
STEMCELL Technologies NeuroCult SM1 Without Vitamin A
Código de producto. 17118291
Encompass
Change view
Click to view available options
Cantidad:
10 mL
Tamaño de la unidad:
10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
17118291 10 mL 10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17118291 Proveedor STEMCELL Technologies N.º de proveedor 05731
Serum-free neural supplement without vitamin A
  • NeuroCult™ SM1 Without Vitamin A is a serum-free culture supplement, based on the published B27 formulation, with vitamin A removed
  • Vitamin A, also known as retinol, induces the differentiation of neural stem cells
  • In combination with a basal medium of choice, NeuroCult™ SM1 Without Vitamin A can be used for primary and pluripotent stem cell (PSC)-derived neural progenitor cell expansion and in customized differentiation protocols where vitamin A is not desired
Especificaciones
Tipo de célula Neural Stem and Progenitor Cells, Neurons, Pluripotent Stem Cells
Para utilizar con (aplicación) Cell Culture, Differentiation, Expansion, Maintenance
Cantidad 10 mL