Learn More
Tocris Bioscience™ NDT 9513727
GREENER_CHOICE
Código de producto. 18013792
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18013792 10 mg 10 miligramos
18025782 50 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18013792 Proveedor Tocris Bioscience™ N.º de proveedor 3333/10
Potent, selective human C5a receptor negative allosteric modulator (NAM)
Potent C5a receptor (C5aR) negative allosteric modulator (NAM) (IC50 = 11.6 nM); selective for human C5aR (IC50 values for rat and mouse C5aR are > 10 μM). Exhibits inverse agonist pharmacokinetics. Blocks degranulation response in C5a-stimulated U937 cells (IC50 = 7.1 nM). Exhibits selectivity for primate and gerbil receptor over other species. Orally bioavailable.
TRUSTED_SUSTAINABILITY
CAS 439571-48-9
Fórmula molecular C36H35N3O4
Especificaciones
Nombre del producto químico o material NDT 9513727
Cantidad 10 mg
Formula Weight (peso de la fórmula) 573.68
Pureza >98%
Peso molecular 573.693