Learn More
Tocris Bioscience™ NBQX disodium salt
Potent AMPA antagonist; more water soluble form of NBQX (Cat. No. 0373)
Marca: Tocris Bioscience™ 1044/10
Descripción
NBQX disodium salt is a selective and competitive AMPA and kainate receptor antagonist (IC50 = 0.15 μM and 4.8 μM, respectively). NBQX blocks the antidepressant effects of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529), decreases mTOR and BDNF levels. NBQX is neuroprotective, anticonvulsant, antinociceptive and active in vivo.NBQX disodium salt is a more water-soluble form of NBQX (Cat. No. 0373).
Especificaciones
| 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt | |
| >98% | |
| Soluble to 100mM in water |
| 479347-86-9 | |
| 10 mg |
For Research Use Only