Learn More
Tocris Bioscience™ (±)-NBI 74330
Descripción
Potent and selective CXCR3 antagonist; potently inhibits 125I-CXCL10 binding to CXCR3 (pKi = 8.13). Inhibits calcium mobilization in response to CXCL11 and CXCL10 in RBL cells. Exhibits no significant effect on chemotaxis induced by CXCR4 or CCR7. Displays a five-fold greater affinity for CXCR3 than (±)-AMG 487 .
This product is racemic.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (±)-NBI 74330 |
| CAS | 473722-68-8 |
| Cantidad | 10 mg |
| Fórmula molecular | C32H27F4N5O3 |
| Formula Weight (peso de la fórmula) | 605.58 |
| Peso molecular | 605.594 |