Learn More
Tocris Bioscience™ NAV 2729
Selective ARF6 inhibitor
Marca: Tocris Bioscience™ 5986/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NAV 2729 is a selective ARF6 inhibitor (IC50 = 1.0 μM). Selective for ARF6 over other ARF family members and small GTPases. Inhibits ARF6 activation in uveal melanoma cells in vitro and blocks downstream signaling pathways of oncogenic GNAQ. Reduces tumorigenesis in mouse model of uveal melanoma.Especificaciones
| Yellow | |
| 419547-11-8 | |
| 456.88g/mol | |
| Store at 4°C | |
| 50 mg |
| ARF GTPase Inhibitors | |
| C25H17ClN4O3 | |
| Soluble to 100mM in DMSO | |
| Blocks downstream signaling pathways |
Título del producto
For Research Use Only