Learn More
Tocris Bioscience™ Napabucasin
Descripción
Napabucasin is a STAT3 inhibitor. Blocks spherogenesis of cancer stem cells (CSC), but not hematopoietic stem cells. Inhibits PaCa-2 xenograft tumor growth in mice, even after cessation of treatment, and decreases population of CSCs in PaCa-2 tumors from treated mice. Also blocks spleen and liver metastases of colon cancer cells in a mouse model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-Acetylnaphtho[2,3-b]furan-4,9-dione |
| CAS | 83280-65-3 |
| Cantidad | 10 mg |
| Objetivo | STAT Inhibitors |
| Fórmula molecular | C14H8O4 |
| Formula Weight (peso de la fórmula) | 240.21 |
| Pureza | >98% |