missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Nanog Polyclonal Antibody, PeproTech®, Invitrogen™

Código de producto. 17805583
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17805583 100 μg 100 microgramos
17845283 50 μg 50 microgramos
17825273 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17805583 Proveedor PeproTech N.º de proveedor 500P236100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: SVDPACPQSL PCFEASDCKE SSPMPVICGP EENYPSLQMS SAEMPHTETV SPLPSSMDLL IQDSPDSSTS PKGKQPTSAE NSVAKKEDKV PVKKQKTRTV FSSTQLCVLN DRFQRQKYLS LQQMQELSNI LNLSYKQVKT WFQNQRMKSK RWQKNNWPKN SNGVTQKASA PTYPSLYSSY HQGCLVNPTG NLPMWSNQTW NNSTWSNQTQ NIQSWSNHSW NTQTWCTQSW NNQAWNSPFY NCGEESLQSC MQFQPNSPAS DLEAALEAAG EGLNVIQQTT RYFSTPQTMD LFLNYSMNMQ PEDV. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Nanog. Anti-Human Nanog-specific antibody was purified by affinity chromatography employing an immobilized Human Nanog matrix. Sandwich ELISA: To detect Human Nanog by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Nanog (500-P236Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Nanog. Western Blot: To detect Human Nanog by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Nanog is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

NANOG (Nanog homeobox) is a divergent homeodomain protein that directs pluripotency and differentiation of undifferentiated embryonic stem cells. NANOG mRNA is present in pluripotent mouse and human cell lines, and absent from differentiated cells. Human NANOG protein shares 52% overall amino acid identity with the mouse protein, and 85% identity in the homeodomain. Human NANOG maps to gene locus 12p13.31, while the mouse NANOG maps to gene loci 6 F2. Murine embryonic NANOG expression is detected in the inner cell mass of the blastocyst. Research studies have shown that high levels of human NANOG expression in the undifferentiated N-Tera embryonal carcinoma cell line. Further, NANOG is a transcription regulator involved in inner cell mass and embryonic stem (ES) cells proliferation and self-renewal. The role of NANOG in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 (also known as Oct-4), another germline-specific transcription factor, and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. Experiments have demonstrated that iPS cells could be generated using expression plasmids expressing NANOG, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. When overexpressed, NANOG promotes cells to enter into S phase and proliferation. Diseases associated with dysfunction in the NANOG protein include tetracarcinoma and germ cell/embryonal cancer.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Nanog
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica NANOG
N.º de referencia del gen Q9H9S0
Alias de gen 2410002E02Rik; early embryo specific expression NK family; early embryo specific expression NK-type homeobox protein; Ecat4; ENK; ES cell-associated protein 4; FLJ12581; HGNC:20857; hNanog; Homeobox protein NANOG; homeobox transcription factor Nanog; homeobox transcription factor Nanog-delta 48; Nanog; Nanog homeobox
Símbolos de los genes NANOG
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human Nanog.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79923
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.