Learn More
Tocris Bioscience™ Nalmefene hydrochloride
Descripción
Opioid receptor antagonist; displays a longer half life than Naloxone . Exhibits neuroprotective effects after traumatic brain injury in the rat.
Deuterated analog also available.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Nalmefene hydrochloride |
| CAS | 58895-64-0 |
| Cantidad | 50 mg |
| Fórmula molecular | C21H26ClNO3 |
| Formula Weight (peso de la fórmula) | 375.89 |
| Pureza | >99% |
| Peso molecular | 375.893 |