Learn More
Naftopidil, MedChemExpress
Código de producto. 16478468
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 g
10 g
Tamaño de la unidad:
10 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16478468 10 g 10 g
16430908 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16478468 Proveedor Medchem Express N.º de proveedor HYB0391
MedChemExpress Naftopidil (KT-611) is is a selective alpha1-adrenoceptor antagonist, with Kis of 3.7 nM, 20 nM and 1.2 nM for the cloned human α1a-, α1b- and α1d-adrenoceptor subtypes, respectively. Naftopidil has antiproliferative effects. Naftopidil can be used for the research of prostate hyperplasia.
Naftopidil (KT-611) is is a selective alpha1-adrenoceptor antagonist, with Kis of 3.7 nM, 20 nM and 1.2 nM for the cloned human α1a-, α1b- and α1d-adrenoceptor subtypes, respectively. Naftopidil has antiproliferative effects. Naftopidil can be used for the research of prostate hyperplasia.
Especificaciones
Nombre del producto químico o material Naftopidil
CAS 57149-07-2
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H28N2O3
Cantidad 10 g
Sinónimo KT-611 BM-15275
Información de solubilidad DMSO : 33.33 mg/mL (84.92 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.25 mM; Need ultrasonic)
SMILES OC(CN1CCN(C(C=CC=C2)=C2OC)CC1)COC3=CC=CC4=CC=CC=C34
Peso molecular (g/mol) 392.49
Formula Weight (peso de la fórmula) 392.49
Porcentaje de pureza 98.97%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation