Learn More
NaDC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16864705 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16864705 20 μL 20 microlitros
16854705 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16864705 Proveedor Proteintech N.º de proveedor 261841AP20UL
Rabbit Polyclonal Antibody
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
Especificaciones
Antígeno NaDC3
Aplicaciones Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.48 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SLC13A3
N.º de referencia del gen Q8WWT9, Q91Y63
Alias de gen hNaDC3, NaDC 3, NADC3, SDCT2, SLC13A3
Símbolos de los genes SLC13A3
Especie del huésped Rabbit
Inmunógeno NaDC3 Fusion Protein Ag24182
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 114644, 64849
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG