Learn More
Tocris Bioscience™ NADA-green
Descripción
NADA-green is a fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. Incorporated into bacterial cell walls during synthesis. Results in strong peripheral and septal labeling of taxonomically diverse bacterial cell populations without affecting growth rate. Excitation/emission λ ~450/555 nm.
please note that the spectral data is solvent dependant
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-[(7-Nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-alanine hydrochloride |
| CAS | 2253733-11-6 |
| Cantidad | 5 mg |
| Fórmula molecular | C9H9N5O5.HCl |
| Formula Weight (peso de la fórmula) | 303.66 |
| Pureza | 98% |