Learn More
STEMCELL Technologies N2 Supplement-A
Código de producto. 17119241
Encompass
Change view
Click to view available options
Cantidad:
5 mL
Tamaño de la unidad:
5 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
17119241 5 mL 5 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17119241 Proveedor STEMCELL Technologies N.º de proveedor 07152
For neural and pancreatic differentiation of mouse and human ES and iPS cells
  • N2 Supplement-A, containing iron-rich human transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural and pancreatic-like cell types
  • Different neuronal subtypes can be generated when human ES/iPS cell-derived neural progenitor cells are cultured in BrainPhys™ Neuronal Medium (Catalog #05790) supplemented with N2 Supplement-A, NeuroCult™ SM1 Neuronal Supplement (Catalog #05711), and other factors
  • N2 Supplement-A is provided as a 100X stock solution
  • N2 Supplement-A is available for individual sale or as a component of the BrainPhys™ Neuronal Medium N2-A & SM1 Kit (Catalog #05793)
Especificaciones
Tipo de célula Endoderm, PSC-Derived, Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pancreatic Cells, Pluripotent Stem Cells
Para utilizar con (aplicación) Cell Culture, Differentiation
Cantidad 5 mL