missing translation for 'onlineSavingsMsg'
Learn More

N-[(Trimethylsilyl)methyl]benzylamine, 95%

Product Code. 15421669
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421669 5 g 5g
15431669 25 g 25g
15441669 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421669 Supplier Thermo Scientific Alfa Aesar Supplier No. H64450.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53215-95-5 | C11H19NSi | 193.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53215-95-5
Molecular Formula C11H19NSi
Molecular Weight (g/mol) 193.37
MDL Number MFCD00012192
InChI Key WECLUYCAWLJMKM-UHFFFAOYSA-N
Synonym n-trimethylsilylmethyl benzylamine, n-trimethylsilyl methyl benzylamine, benzyl trimethylsilyl methyl amine, n-benzyl-1-trimethylsilyl methylamine, 1-phenyl-n-trimethylsilylmethyl methanamine, benzyl trimethylsilylmethyl amine, n-benzyl-1-trimethylsilyl methanamine, benzenemethanamine, n-trimethylsilyl methyl, n-benzylaminomethyltrimethylsilane, n-benzyl-n-trimethylsilyl methyl amine
PubChem CID 353458
SMILES C[Si](C)(C)CNCC1=CC=CC=C1

Specifications

Color Yellow
Density 0.88 g/mL
Boiling Point 89°C to 90°C (5 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.496
Quantity 5 g
Beilstein 2802488
IUPAC Name 1-phenyl-N-(trimethylsilylmethyl)methanamine
Formula Weight 193.36
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine
Show More Show Less
compliance-icons
Product Identifier
  • N-[(Trimethylsilyl)methyl]benzylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N-[(Trimethylsilyl)methyl]benzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.