Learn More
Ácido N-metiliminodiacético, 99 %, Thermo Scientific Chemicals
Código de producto. 11347755
Cambiar vista
Click to view available options
Cantidad:
5 g
25 g
Tamaño de la unidad:
25 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11347755 25 g 25 g
11337755 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11347755 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor L02589.14
Solicitar formato a granel o personalizado
CAS: 4408-64-4 | C5H9NO4 | 147.13 g/mol
N-Methyliminodiacetic acid is used to prepare bis-(1H-benzoimidazol-2-ylmethyl)-methyl-amine by condensation with benzene-1,2-diamine. It is involved in Suzuki cross-coupling with N-methyliminodiacetic acid (MIDA) boronates. It acts as a reagent utilized for protecting boronic acids and as a chelating agent for various metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
el ácido N-metiliminodiacético se utiliza para preparar la bis-(1H-benzoimidazol-2-ilmetil)-metil-amina por condensación con benceno-1,2-diamina. Participa en el acoplamiento cruzado de Suzuki con boronatos de ácido N-metiliminodiacético (MIDA). Actúa como un reactivo que se utiliza para proteger los ácidos borónicos y como agente quelante para diversos metales.
Solubilidad
Soluble en agua. Ligeramente soluble en dimetilsulfóxido.
Notas
Incompatible con bases fuertes.
TRUSTED_SUSTAINABILITY
CAS 4408-64-4
Fórmula molecular C5H9NO4
Peso molecular (g/mol) 147.13
Número MDL MFCD00004284
Clave InChI XWSGEVNYFYKXCP-UHFFFAOYSA-N
Sinónimo n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
PubChem CID 20441
Nombre IUPAC ácido 2-[carboximetil(metil)amino]acético
SMILES CN(CC(=O)O)CC(=O)O
Especificaciones
Nombre del producto químico o material N-Methyliminodiacetic acid
Punto de fusión ∽220°C (decomposition)
Cantidad 25 g
Beilstein 1765449
Información de solubilidad Soluble in water. Slightly soluble in dimethyl sulfoxide.
Formula Weight (peso de la fórmula) 147.13
Porcentaje de pureza 99%