Learn More
Tocris Bioscience™ N-methylated pomalidomide
Descripción
N-methylated pomalidomide is a functionalized cereblon ligand for PROTAC® research and development. Can be used to generate negative controls for Pomalidomide (Cat. No. 6302)-based Degraders. Does not bind to cereblon. Part of a range of functionalized tool molecules for PROTAC R&D. Can also be used as a Pomalidomide negative control.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Amino-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 1352827-50-9 |
| Cantidad | 50 mg |
| Objetivo | Active Degrader Negative Controls |
| Fórmula molecular | C14H13N3O4 |
| Pureza | 0.95 |