Learn More
R&D Systems™ N-glycan Labeling and Detection Kit
Specifications
Specifications
| Cantidad | 1 Kit |
| Requisitos de almacenamiento | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Objetivo | Multi-Species |