Learn More
N-Boc-3-dimetilamino-L-alanina, 97 %, Thermo Scientific Chemicals
Descripción
N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-3-dimetilamino-L-alanina se utiliza como un intermediario de síntesis química orgánica.
Notas
Estable en las condiciones de almacenamiento recomendadas. Almacenar lejos de agentes oxidantes.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-Boc-3-dimethylamino-L-alanine |
| Cantidad | 250 mg |
| Formula Weight (peso de la fórmula) | 232.28 |
| Porcentaje de pureza | 97% |