Learn More
Tocris Bioscience™ N-Acetylpuromycin
Descripción
N-Acetylpuromycin downregulates SnoN and Ski protein expression; promotes TGF-β signaling, independently of MAPK activation. Does not bind ribosomes or block protein synthesis.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3′-[[(2S)-2-(Acetylamino)-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3′-deoxy-N,N-dimethyladenosine |
| CAS | 22852-13-7 |
| Cantidad | 10 mg |
| Objetivo | Additional TGF-beta Receptor Products |
| Fórmula molecular | C24H31N7O6 |
| Formula Weight (peso de la fórmula) | 513.55 |
| Pureza | >98% |