Learn More
MZ 1, MedChemExpress
Código de producto. 16491079
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16491079 1 mg 1 miligramo
16486827 5 mg 5 miligramos
16474128 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16491079 Proveedor Medchem Express N.º de proveedor HY107425
MedChemExpress MZ 1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4. MZ 1 potently and rapidly induces reversible, long-lasting, and selective removal of BRD4 over BRD2 and BRD3. Kds of 382/120, 119/115, and 307/228 nM for BRD4 BD1/2, BRD3 BD1/2, and BRD2 BD1/2, respectively.
MZ 1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4. MZ 1 potently and rapidly induces reversible, long-lasting, and selective removal of BRD4 over BRD2 and BRD3. Kds of 382/120, 119/115, and 307/228 nM for BRD4 BD1/2, BRD3 BD1/2, and BRD2 BD1/2, respectively.
Especificaciones
Nombre del producto químico o material MZ 1
CAS 1797406-69-9
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C49H60ClN9O8S2
Cantidad 1 mg
Información de solubilidad DMSO : 100 mg/mL (99.74 mM; Need ultrasonic)
SMILES CC1=C(C)SC2=C1C(C3=CC=C(Cl)C=C3)=N[C@@H](CC(NCCOCCOCCOCC(N[C@@H](C(C)(C)C)C(N4[C@@H](C[C@@H](O)C4)C(NCC5=CC=C(C6=C(N=CS6)C)C=C5)=O)=O)=O)=O)C7=NN=C(C)N27
Peso molecular (g/mol) 1002.64
Formula Weight (peso de la fórmula) 1002.64
Porcentaje de pureza 95.21%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death