Learn More
MyoD1 Ab-2 Mouse Monoclonal Antibody, Epredia™
Código de producto. p-4553315 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
Concentración:
1mg/mL
200μg/mL
Tamaño de la unidad:
1 ML
100 microlitros
200 microlitros
500 microlitros
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Concentración unitSize
12600787 100 μL 200μg/mL 100 microlitros
12610787 500 μL 200μg/mL 500 microlitros
12690777 1 mL 200μg/mL 1 ML
12620787 100 μL 1mg/mL 100 microlitros
12630787 200 μL 1mg/mL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 12600787 Proveedor Epredia N.º de proveedor MS278P0
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Deliver accurate, reproducible results with the Epredia™ MyoD1 Ab-2, Mouse Monoclonal Antibody.
Research Areas: Muscle Markers; MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.
Host Species: Mouse
Clone: 5.2F
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Chicken. Others not known.
Epitope: aa 3-56
Immunogen: Recombinant mouse MyoD1 protein
Molecular Weight: 45kDa
Positive Control: SW80 cells
Cellular Localization: Nuclear
Recommended for:
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
Note:
USA: RUO; Int'l: RUO
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno MyoD1 Ab-2
Aplicaciones Immunofluorescence, Immunoprecipitation
Clasificación Monoclonal
Clon 5.2F
Concentración 200μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Recombinant mouse MyoD1 protein
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Muscle Biology
Primario o secundario Primary
Especies diana Chicken, Human, Mouse, Rat
Formulario Concentrated
Isotype IgG2a κ