missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTMR3 Polyclonal Antibody

Descripción
Immunogen sequence: KWQEHRRSLE LSSLAGPGED PLSADSLGKP TRVPGGAELS VAAGVAEGQM ENILQEATKE ESGVEEPAHR AGIEIQEGKE DPLLEKESRR KTPEASAIGL HQDPELGDAA LRSHLDMSWP LFSQGISEQQ SGLSVLLSSL QVPPRGEDSL EVPVEQFRIE EIAEGREEAV L.
This gene encodes a member of the myotubularin dual specificity protein phosphatase gene family. The encoded protein is structurally similar to myotubularin but in addition contains a FYVE domain and an N-terminal PH-GRAM domain. The protein can self-associate and also form heteromers with another myotubularin related protein. The protein binds to phosphoinositide lipids through the PH-GRAM domain, and can hydrolyze phosphatidylinositol(3)-phosphate and phosphatidylinositol(3,5)-bisphosphate in vitro. The encoded protein has been observed to have a perinuclear, possibly membrane-bound, distribution in cells, but it has also been found free in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | MTMR3 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.64 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| génica | MTMR3 |
| N.º de referencia del gen | Q13615 |
| Alias de gen | 1700092A20Rik; AI255150; AW557713; FYVE (Fab1 YGLO23 Vsp27 EEA1 domain) dual-specificity protein phosphatase; FYVE domain-containing dual specificity protein phosphatase 1; FYVE-DSP1; KIAA0371; mKIAA0371; MTMR3; myotubularin related protein 3; myotubularin-related protein 3; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; ZFYVE10; zinc finger FYVE domain-containing protein 10; zinc finger, FYVE domain containing 10 |
| Símbolos de los genes | MTMR3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?