missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/MSK2 Rabbit anti-Human, Clone: 1013F, R&D Systems™

Rabbit Monoclonal Antibody
Marca: R&D Systems MAB25181-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry.Especificaciones
| MSK1/MSK2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB | |
| Phosphopeptide containing the human MSK1(S376) site. Accession # O75582 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 1013F | |
| Immunocytochemistry 1-25 ug/mL | |
| O75582 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. | |
| Human | |
| Purified |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?