Learn More
Tocris Bioscience™ MS 28
Descripción
MS 28 is a cyclinD1 Degrader (PROTAC™) (DC50 = 950 nM; Dmax = 90% in vitro). Also degrades CDK4, CDK6 and cyclin D3 after 4 hours. Degrades cyclin D1 and cyclin D3 by recruiting the CDK4/6 as a 'bridge'. Comprises a CDK4/6 binding moiety joined by a linker to a VHL ligand. Suppresses NCI-H2110 and Calu-1 cancer cell growth at 1 μM.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2S,4R)-1-((S)-2-(7-(4-(6-((6-Acetyl-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino)pyridin-3-yl)piperazin-1-yl)-7-oxoheptanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2229974-73-4 |
| Cantidad | 5 mg |
| Objetivo | Active Degraders |
| Fórmula molecular | C53H67N11O7S |
| Pureza | 0.98 |