Learn More
Tocris Bioscience™ MS 143
Descripción
MS 143 is a potent and selective protein kinase B (AKT) Degrader (PROTAC™) (DC50 = 46 nM, Dmax = 100 % at 24h in PC3 cells). MS 143 suppresses growth in prostate cancer PC3 cells; suppresses tumor growth and reduces AKT protein levels in a mouse PC3 cell xenograft model.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Amino-N-((S)-1-(4-chlorophenyl)-3-(4-(12-(((S)-1-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-12-oxododecanoyl)piperazin-1-yl)propyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4 |
| CAS | 2376137-41-4 |
| Cantidad | 5 mg |
| Objetivo | Active Degraders |
| Fórmula molecular | C59H81ClN12O6S |
| Pureza | 97% |