Learn More
Tocris Bioscience™ MS 023 dihydrochloride
Descripción
MS 023 dihydrochloride is a potent and selective type I PRMT inhibitor (IC50 values are 8, 8, 30, 83 and 119 nM for PRMT6, PRMT8, PRMT1, PRMT4 and PRMT3, respectively). Exhibits no significant activity against type II and III PRMTs, PKMTs, DNMTs, KDMs and reader proteins. Reduces global arginine asymmetric dimethylation levels in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N1-Methyl-N1-[[4-[4-(1-methylethoxy)phenyl]-1H-pyrrol-3-yl]methyl]-1,2-ethanediamine dihydrochloride |
| CAS | 1992047-64-9 |
| Cantidad | 10 mg |
| Objetivo | Histone Methyltransferase Inhibitors |
| Fórmula molecular | C17H25N3O.2HCl |
| Formula Weight (peso de la fórmula) | 360.32 |
| Pureza | >98% |