Learn More
Tocris Bioscience™ MRT 67307 dihydrochloride
Descripción
MRT 67307 dihydrochloride is a salt inducible kinase (SIK) inhibitor (IC50 values are 67, 250 and 430 nM for SIK2, SIK1 and SIK3 respectively). Also inhibits TBK1, MARK1-4, IKKε and NUAK1 (IC50 values are 19, 27-52, 160 and 230 nM respectively). Has no effect on IKKα or IKKβ. Induces IL-10 secretion and inhibits TNF-α and IL-6 secretion in bacterial LPS-stimulated macrophages. Also enhances IL-1-induced activation of NFκB-dependent gene transcription in mouse embryonic fibroblast (MEF) cells. Inhibits autophagy.
Especificaciones
Especificaciones
| CAS | 1781882-89-0 |
| Cantidad | 10 mg |
| Tipo de producto | MRT 67307 dihydrochloride |
| Pureza | >98% |
| Peso molecular | 537.52 |
| Inhibidores | MRT 67307 dihydrochloride |
Para uso exclusivo en investigación