Learn More
Tocris Bioscience™ MRS 2690
Descripción
MRS 2690 is a potent P2Y14 receptor agonist (EC50 = 49 nM) that displays 7-fold higher potency than UDP-glucose. This product is typically reconstituted in water.
Especificaciones
Especificaciones
| Nombre del producto químico o material | MRS 2690 |
| CAS | 15039-58-4 |
| Cantidad | 1 mg |
| Fórmula molecular | C15H22N2Na2O16P2S |
| Formula Weight (peso de la fórmula) | 626.33 |
| Pureza | >98% |