Learn More
Tocris Bioscience™ MRS 2578
Descripción
MRS 2578 is a selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
Especificaciones
Especificaciones
| Cantidad | 10 mg |
| Tipo de producto | MRS 2578 |
| Peso molecular | 472.66 |
| Inhibidores | MRS 2578 |
Para uso exclusivo en investigación