missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL48 Polyclonal Antibody

Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 6p.
Especificaciones
Especificaciones
| Antígeno | MRPL48 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | MRPL48 |
| N.º de referencia del gen | 0 |
| Alias de gen | 1810030E20Rik; 2610028L11Rik; 39S ribosomal protein L48, mitochondrial; CGI-118; D4Ertd786e; HSPC290; L48mt; MGC13323; mitochondrial 39S ribosomal protein L48-like protein; Mitochondrial large ribosomal subunit protein mL48; mitochondrial ribosomal protein L48; Mrpl48; MRP-L48; Unknown (protein for MGC:133957) |
| Símbolos de los genes | MRPL48 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?