missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL32 Polyclonal Antibody
Descripción
The antibody detects endogenous levels of total MRPL32 protein.
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-L32 (mitochondrial ribosomal protein L32), also known as HSPC283, is a 188 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 39S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. The gene encoding MRP-L32 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Especificaciones
Especificaciones
| Antígeno | MRPL32 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | MRPL32 |
| N.º de referencia del gen | Q9BYC8 |
| Alias de gen | 0610033O15Rik; 39S ribosomal protein L32, mitochondrial; bMRP-59b; Heart-expressed gene 1 protein; Heg1; HSPC283; L32mt; Mitochondrial large ribosomal subunit protein bL32m; mitochondrial ribosomal protein L32; MRPL32; MRP-L32 |
| Símbolos de los genes | MRPL32 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?