Learn More
Tocris Bioscience™ MPro N3
Descripción
MPro N3 is a coronavirus main protease (Mpro) inhibitor (respective IC50 values are 2.7, 4 and 8.8 μM for MHV-A29, HCoV-229E and FOPV replication in vitro). Inhibits viral cell infection in an MHV plaque reduction assay. Also inhibits SARS-CoV-2 viral cell entry in a plaque reduction assay (IC50 = 16.8 μM).
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[(5-Methyl-3-isoxazolyl)carbonyl]-L-alanyl-L-valyl-N-[(1 S,2 E)-4-oxo-1-[[(3 S)-2-oxo-3-pyrrolidinyl]methyl]-4-(phenylmethoxy)-2-buten-1-yl]-L-leucinamide |
| CAS | 884650-98-0 |
| Cantidad | 25 mg |
| Objetivo | Additional Synthetic Protease Inhibitors |
| Fórmula molecular | C35H48N6O8 |
| Pureza | 0.98 |