Learn More
STEMCELL Technologies Kit de ELISA para TNF-alfa de ratón
Descripción
- The Mouse Tumor Necrosis Factor-alpha (TNF-α) ELISA Kit is designed for the quantitative detection and measurement of mouse TNF-α in biological fluids such as serum, plasma, and cell culture supernatants
- TNF-α is produced by activated T cells and macrophages in response to inflammation or infection
- It is also produced by B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Especificaciones
| Descripción | B Cells; Cardiomyocytes, PSC-Derived; Dendritic Cells; Endothelial Cells; Granulocytes and Subsets; Macrophages; Mouse Embryonic Fibroblasts; Myogenic Stem and Progenitor Cells; NK Cells; T Cells |
| Para utilizar con (aplicación) | ELISA |
| Cantidad | 2 placas |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?