missing translation for 'onlineSavingsMsg'
Learn More

Mouse TER-119 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Código de producto. 18781198
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18781198 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18781198 Proveedor R&D Systems N.º de proveedor FAB1125U100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody

TER-119 Monoclonal specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TER-119
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon TER-119
Concentración 0.2 mg/mL
Conjugado Alexa Fluor 350
Dilución Flow Cytometry 0.25-1 ug/10^6 cells
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Alias de gen Ly76, Ly-76, TER119
Símbolos de los genes Ly76
Especie del huésped Rat
Inmunógeno C57BL/6 mouse day-14 fetal liver cells
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 104231
Especificidad de la prueba Detects mouse TER-119 in Western blots. This antibody has been shown to react with cells of the erythroid lineage in embryonic yolk sac, fetal liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs (Ikuta, K. et al. (1990) Cell 62:863). TER-119 Antigen is a molecule associated with cell-surface glycophorin A (Kina, T. et al. (2000) Br. J. Haematol. 109:280). The antibody has been shown to react with erythroid cells at differentiation stages from early proerythroblast to mature erythrocyte (Kina, T. et al. (2000) Br. J. Haematol. 109:280).
Especies diana Mouse
Contenido y almacenamiento Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.